Search
Hoechst 33342 nucleic acid stain is a popular cell-permeant nuclear counterstain that emits blue fluorescence when bound to dsDNA. It is also available as a solution.
| Hoechst 33342 | |
|---|---|
| Excitation/Emission (nm) | 350/461 |
| Standard filter set | DAPI |
| EVOS Light Cube | DAPI |
| Storage conditions | 2–6°C or ≤–20°C |
Cells stained with Hoechst 33342 and imaged with the Thermo Scientific CellInsight High-Content System.