Search
Learn the fundamentals of protein expression using the Gibco Expi293 Expression Systems with detailed overviews of the unique system components and key workflow steps for which optimal techniques will help maximize results. This protocol guide includes protocol videos, a protein expression protocol calculator, required material checklists, visual step-by-step guides, and helpful tips.
At the end of this protocol guide, you should be able to:
The Expi293 expression systems are high-yield transient expression systems based on suspension-adapted Human Embryonic Kidney (HEK) cells. Each Expi293 expression system starter kit provides cells, culture medium, and reagents to express a total of 1 liter production volume. All of the system components work together to allow access to recombinant 293-derived proteins in just 5-7 days, with a 2-10 fold increase in protein yields compared to previous generation transient expression systems.
The Expi293 expression systems offer four Expi293F cell lines options as well as one labeling kit option to suit a broad range of applications and proteins. We recommend to:
Expi293 Expression System Kit (Cat. No. A14635)
Components |
Amount |
Cat. No. |
Storage |
| Expi293F Cells* (1 × 107 cells/mL) | 2 x 1 mL |
Liquid nitrogen** |
|
| Expi293 Expression Medium | 1 L |
2°C to 8°C Protect from light |
|
ExpiFectamine 293 Transfection Kit:
|
1 kit for 1 L of culture |
||
100 mL |
|||
pRABBIT IgG IRES-EmGFP Positive Control Vector (at 1 mg/mL in TE‡ Buffer, pH 8.0) |
150 µg |
−20°C |
*Cells are cryopreserved in cryopreservation medium consisting of 42.5% fresh Expi293 Expression Medium, 50.0% conditioned medium and 7.5% DMSO.
**Store the frozen cells in liquid nitrogen immediately upon receipt and until ready to use. Do not store the cells at −80°C.
†Opti-MEM I Reduced Serum Medium is a serum-free reagent.
‡TE buffer, pH 8.0: 10 mM Tris-HCl, 1 mM EDTA, pH 8.0.
Expi293 Met (-) Protein Labeling Kit (Cat. No. A41249)
Components |
Amount |
Cat. No. |
Storage |
ExpiFectamine 293 Met (-) Transfection Kit
|
1 kit for 1 L culture |
2°C to 8°C Protect from light. |
|
1 L |
Expi293 GnTI- Expression System Kit (Cat. No. A39250)
Components |
Amount |
Cat. No. |
Storage |
Expi293F GnTI- Cells* (1 × 107 cells/mL) |
1 mL |
Liquid nitrogen** |
|
1 L |
2°C to 8°C Protect from light |
||
ExpiFectamine 293 Transfection Kit:
|
1 kit for 1 L of culture |
||
100 mL |
|||
pRABBIT IgG IRES-EmGFP Positive Control Vector (at 1 mg/mL in TE† Buffer, pH 8.0) |
150 µg |
−20°C |
|
1 Kit |
−20°C |
*Cells are cryopreserved in cryopreservation medium consisting of 42.5% fresh Expi293 Expression Medium, 50.0% conditioned medium and 7.5% DMSO.
**Store the frozen cells in liquid nitrogen immediately upon receipt and until ready to use. Do not store the cells at −80°C.
†TE buffer, pH 8.0: 10 mM Tris-HCl, 1 mM EDTA, pH 8.0.
Expi293 Inducible Expression System Kit (Cat. No. A39251)
Components |
Amount |
Cat. No. |
Storage |
Expi293F Inducible Cells* (1 × 107 cells/mL) |
1 mL |
Liquid nitrogen** |
|
1 L |
2°C to 8°C Protect from light |
||
ExpiFectamine 293 Transfection Kit:
|
1 kit for 1 L of culture |
||
100 mL |
|||
pRABBIT IgG IRES-EmGFP Positive Control Vector (at 1 mg/mL in TE† Buffer, pH 8.0) |
150 µg |
−20°C |
|
20 µg |
|||
500 mg |
Protect from light and moisture Room temperature |
*Cells are cryopreserved in cryopreservation medium consisting of 42.5% fresh Expi293 Expression Medium, 50.0% conditioned medium and 7.5% DMSO.
**Store the frozen cells in liquid nitrogen immediately upon receipt and until ready to use. Do not store the cells at −80°C.
†TE buffer, pH 8.0: 10 mM Tris-HCl, 1 mM EDTA, pH 8.0.
‡Not included in Cat. No. A41275.
Expi293 Inducible GnTI- Expression System Kit (Cat. No. A39252)
Components |
Amount |
Cat. No. |
Storage |
Expi293F Inducible GnTI- Cells* (1 × 107 cells/mL) |
1 mL |
Liquid nitrogen** |
|
1 L |
2°C to 8°C Protect from light |
||
ExpiFectamine 293 Transfection Kit:
|
1 kit for 1 L of culture |
||
100 mL |
|||
pRABBIT IgG IRES-EmGFP Positive Control Vector (at 1 mg/mL in TE† Buffer, pH 8.0) |
150 µg |
−20°C |
|
20 µg |
|||
500 mg |
Protect from light and moisture Room temperature |
||
1 kit |
−20°C |
*Cells are cryopreserved in cryopreservation medium consisting of 42.5% fresh Expi293 Expression Medium, 50.0% conditioned medium and 7.5% DMSO.
**Store the frozen cells in liquid nitrogen immediately upon receipt and until ready to use. Do not store the cells at −80°C.
†TE buffer, pH 8.0: 10 mM Tris-HCl, 1 mM EDTA, pH 8.0.
‡Not included in Cat. No. A41276.
The following checklist provides a concise list of recommended consumables, instruments, and equipment to guide you in starting your protein expression experiment.
Key lab equipment and instruments |
Key lab consumables |
|---|---|
Biosafety cabinet with HEPA filter Incubator(opens in a new tab)/shaker (temperature, humidity and CO2 regulated) Aspiration pump (peristaltic or vacuum) Waste containers and vacuum traps |
Cell culture flasks, vented, non-baffled (125 mL to 5 L) Serological pipettes and pipettors (1 mL to 50 mL) Multichannel pipettors and pipettes (1 µL to 200 µL) Single channel pipettors and pipettes (1 µL to 1 mL) Cell culture media, PBS and sterile H2O Personal protective equipment (Lab coats, safety goggles, disposable gloves, eye wash station) |
General cell handling |
|
|
|
|
|
Guidelines for thawing and storing cells |
|
|
|
|
|
Guidelines for cell maintenance and subculturing |
|
|
|
|
|
|
|
|
|
|
|
Guidelines for inducible cells |
For Expi293F Inducible Cells or Expi293F Inducible GnTI- Cells: |
|
|
|
|
|
|
|
|
Guidelines for media |
|
| IMPORTANT! Expi293 Expression Medium is sensitive to light. For optimal results, use and store media protected from light. | |
| Note: ExpiFectamine 293 Transfection Enhancer 1 can exhibit a slightly yellowish tint. Internal studies show this has no impact on system performance or protein titer. |
In this video, learn more about the best practices for thawing, culturing and banking HEK293 suspension adapted cells, including Expi293F cells and Gibo viral production cells.
Note: the instructions below are applicable to all Expi293F cell lines including Expi293F Cells, Expi293F GnTI- Cells, Expi293F Inducible Cells, and Expi293F Inducible GnTI- Cells.
Thaw cells (Day 1)
|
1. Add 30 mL of pre-warmed Expi293 expression medium to a 125‑mL Erlenmeyer shaker flask 2. Remove the vial of cells from liquid nitrogen and swirl it in a 37°C water bath for 1–2 min to rapidly thaw the cells until only a small amount of ice remains. Note: Do not fully submerge the vial in the water bath. 3. Decontaminate the external surface of the vial by wiping it with 70% ethanol before opening the vial in the BSC or laminar flow hood. |
|
Add cells to medium (Day 1)
|
4. Carefully pipette the entire contents of the vial into the 125 mL Erlenmeyer shake flask containing 30 mL pre‑warmed Expi293 expression medium. |
|
Incubate cells
|
5. Incubate the cells at 37°C on the orbital shaker with ≥80% relative humidity and 8% CO2. |
|
Shaking speed |
19 mm shaking diameter: 125 ± 5 rpm 25 mm shaking diameter: 120 ± 5 rpm 50 mm shaking diameter: 95 ± 5 rpm |
|
Count cells and determine viability (Day 3)
|
6. On Day 3 post-thaw, determine viable cell density and percent cell viability. Cell viability should be ≥90% with a viable cell density >1 × 106 viable cells/mL. 7. For subsequent routine cell culture maintenance, subculture cells every 3 to 4 days when the viable cell density reaches 3 × 106 to 5 × 106 viable cells/mL. |
|
In this video, learn how to produce recombinant proteins with the Gibco Expi293 expression system and achieve gram per liter protein yields in a transient HEK 293 system.
Note: the instructions below are applicable to all Expi293F cell lines including Expi293F Cells, Expi293F GnTI- Cells, Expi293F Inducible Cells, and Expi293F Inducible GnTI- Cells.
Passage cells (Day 1)
|
1. Based on the measured viable cell density, calculate the volume of cell suspension required to seed a new shake flask according to the recommended seeding densities in Table 1 and the recommended culture volumes in Table 2. 2. Transfer the calculated volume of cells to pre-warmed Expi293 Expression Medium in the Erlenmeyer shake flask(s). |
Subculturing and harvest
|
3. Incubate flask(s) on the orbital shaker in the 37°C incubator with ≥80% relative humidity and 8% CO2 until culture(s) reach a density of 3 x 106 to 5 x 106 viable cells per mL. Note: Cells that are subcultured at densities outside of this early log phase growth window may show longer doubling times and lower titers over time. If necessary, modify the initial seeding density to attain the target cell density of 3 x 106 to 5 x 106 viable cells per mL at the subculturing stage. 4. Repeat Steps 1–3 to maintain or expand the cells for transfection. |
Table 1. Recommended seeding densities for routine cell culture maintenance.
Subculture timing |
Recommended seeding density |
For cells ready 3 days post-subculture |
0.4 x 106 to 0.5 x 106 viable cells per mL |
For cells ready 4 days post-subculture |
0.3 x 106 to 0.4 x 106 viable cells per mL |
Table 2. Recommended volumes and shaking speeds for routine cell culture maintenance.
Flask size |
125 mL |
250 mL |
500 mL |
1L |
2 L |
3 L |
Culture volume (mL) |
30–35 |
60–70 |
100–120 |
220–240 |
440–480 |
800–1,000 |
Shaking speed* |
19 mm shaking diameter: 125 ± 5 rpm 25 mm shaking diameter: 120 ± 5 rpm 50 mm shaking diameter: 95 ± 5 rpm |
90 ± 5 rpm 85 ± 5 rpm 80 ± 5 rpm |
||||
Flask type |
Vented, non-baffled |
|||||
For Research Use Only. Not for use in diagnostic procedures.