Search Thermo Fisher Scientific
In this assay, the modified thymidine analog EdU (5-ethynyl-2′-deoxyuridine, a nucleoside analog of thymidine) is efficiently incorporated into newly synthesized DNA and fluorescently labeled with a bright, photostable Alexa Fluor® dye in a fast, highly specific, mild click reaction.
| Reaction components* | Number of plates | ||||
|---|---|---|---|---|---|
| 0.5 | 1 | 2 | 5 | 10 | |
| 1X Click-iT® EdU reaction buffer | 5.1 mL | 10.3 mL | 20.6 mL | 51.5 mL | 103 mL |
| CuSO4 (Component D) | 240 μL | 480 μL | 960 μL | 2.4 mL | 4.8 mL |
| Alexa Fluor® azide (Component B) | 15 μL | 30 μL | 60 μL | 150 μL | 300 μL |
| 1X Click-iT® EdU buffer additive | 600 μL | 1.2 mL | 2.4 mL | 6 mL | 12 mL |
| Total volume | 6 mL | 12 mL | 24 mL | 60 mL | 120 mL |
| *Note: Add the ingredients in the order listed in the table. | |||||
| HCS NuclearMask™ Blue stain | Alexa Fluor® 488 | Alexa Fluor® 555 | Alexa Fluor® 594 | Alexa Fluor® 647 | |
|---|---|---|---|---|---|
| Excitation/Emission (in nm) | 350/461 | 495/519 | 555/565 | 590/615 | 650/670 |
| Standard filter set | DAPI | FITC | RFP | TRITC | Cy®5 |