Search
| Color (Ex/Em *) |
Bead Diameter
| |||
|---|---|---|---|---|
| 15 µm | 10 µm | |||
| Blue (365/415) | F8837 | F8829 | ||
| Blue-green (430/465) | F8838 | F8830 | ||
| Green (450/480) | F21010 | |||
| Yellow-green (505/515) | F8844 | F8836 | ||
| Yellow (515/534) | F21011 | |||
| Orange (540/560) | F8841 | F8833 | ||
| Red-orange (565/580) | F21012 | |||
| Red (580/605) | F8842 | F8834 | ||
| Carmine (580/620) | F21013 | |||
| Crimson (625/645) | F8839 | F8831 | ||
| Scarlet (645/680) | F8843 | |||
| * Fluorescence excitation and emission maxima of the encapsulated dye measured in 2-ethoxyethyl acetate. All FluoSpheres microspheres for blood flow determination are supplied as suspensions containing 1.0 × 106 beads/mL (15 µm microspheres) or 3.6 × 106 beads mL (10 µm microspheres) in a unit size of 10 mL. | ||||
For Research Use Only. Not for use in diagnostic procedures.