Search
Size
| |||
|---|---|---|---|
| Excitation/Emission (nm) * |
0.04 µm
|
0.1 µm
|
1.0 µm
|
| 488/560 | T8864 | T8872 | T8880 |
| 488/605 NeutrAvidin-labeled | T8860 | T8861 | |
| 488/645 | T8883 | ||
| 488/645 Streptavidin-labeled | T10711 | ||
| 633/720 | T8870 | ||
| * All TransFluoSpheres beads are based on carboxylate-modified polystyrene microspheres and are supplied as 0.5 mL of an aqueous suspension containing 2% solids and 2 mM sodium azide, except the TransFluoSpheres beads labeled with NeutrAvidin biotin-binding protein, which are supplied as 0.4 mL of an aqueous suspension containing 1% solids and 2 mM sodium azide, and the streptavidin-labeled beads, which are supplied as 0.4 mL of an aqueous suspension containing 0.5% solids and 2 mM sodium azide. | |||