Tyramide Signal Amplification (TSA) Kits—Table 6.1
Table 6.1 Tyramide Signal Amplification (TSA) Kits.
Tyramide (Ex/Em) * | Peroxidase Conjugate | ||
---|---|---|---|
Anti–Mouse IgG † | Anti–Rabbit IgG † | Streptavidin | |
Alexa Fluor 350 (346/442) | TSA Kit #7 (T20917) | TSA Kit #17 (T20927) | TSA Kit #27 (T20937) |
Alexa Fluor 488 (495/519) | TSA Kit #2 (T20912) | TSA Kit #12 (T20922) | TSA Kit #22 (T20932) |
Oregon Green 488 (496/524) | TSA Kit #29 (T20939) | ||
Alexa Fluor 546 (556/573) | TSA Kit #3 (T20913) | TSA Kit #13 (T20923) | TSA Kit #23 (T20933) |
Alexa Fluor 555 (555/565) | TSA Kit #40 (T30953) | TSA Kit #41 (T30954) | TSA Kit #42 (T30955) |
Alexa Fluor 568 (578/603) | TSA Kit #4 (T20914) | TSA Kit #14 (T20924) | TSA Kit #24 (T20934) |
Alexa Fluor 594 (590/617) | TSA Kit #5 (T20915) | TSA Kit #15 (T20925) | TSA Kit #25 (T20935) |
Alexa Fluor 647 (650/668) | TSA Kit #6 (T20916) | TSA Kit #16 (T20926) | TSA Kit #26 (T20936) |
Biotin-XX (NA) | TSA Kit #1 (T20911) | TSA Kit #11 (T20921) | TSA Kit #21 (T20931) |
*Fluorescence excitation (Ex) and emission (Em) maxima, in nm. † Host = goat. NA = Not applicable. |
For Research Use Only. Not for use in diagnostic procedures.