Alkaline Phosphatase and Horseradish Peroxidase Enzyme Conjugates—Table 7.3
Learn more in The Molecular Probes® Handbook
Table 7.3 Alkaline phosphatase and horseradish peroxidase enzyme conjugates.
Conjugate or Zenon Antibody Labeling Kit |
Enzyme Conjugate
|
|
---|---|---|
Alkaline Phosphatase | Horseradish Peroxidase | |
Antibody (Host) | ||
Anti–mouse IgG (goat) | G21060 | G21040 |
Anti–mouse IgG (γ1) (goat) | A10551 | |
Anti–mouse IgG, IgM (goat) | A10678 | A10677 |
Anti–mouse IgG, IgM, IgA (goat) | A10668 | |
Anti–mouse IgG, F(ab')2 fragment (goat) | F21452 | F21453 |
Anti–mouse IgG (rabbit) | R21455 | |
Anti–rabbit IgG (goat) | G21079 | G21234 |
Anti–rabbit IgG, F(ab')2 fragment (goat) | F21456 | A10547 |
Anti–rat IgG (goat) | A10549 | |
Anti–rat IgG, F(ab')2 fragment (goat) | A10548 | |
Anti–human IgG1 (mouse) | A10649 | A10648 |
Anti–human IgG4 (mouse) | A10660 | A10654 |
Anti–goat IgG (rabbit) | R21458 | R21459 |
Anti–fluorescein/Oregon Green (rabbit) | A21253 | |
Anti–Green Fluorescent Protein (rabbit) | A10260 | |
Zenon Antibody Labeling Kit | ||
Zenon Mouse IgG1 Labeling Kit | Z25054 | |
Zenon Mouse IgG2a Labeling Kit | Z25154 | |
Zenon Mouse IgG2b Labeling Kit | Z25254 | |
Zenon Rabbit IgG Labeling Kit | Z25350 | Z25354 |
Zenon Human IgG Labeling Kit | Z25454 | |
Avidin and Protein G Conjugates | ||
NeutrAvidin biotin-binding protein | A2664 | |
Streptavidin | S921 | S911 |
Protein G | P21041 |
For Research Use Only. Not for use in diagnostic procedures.