Search
| Dyes (Cat. No.) | Structure (Charge) | Optical Response | Selected Applications |
|---|---|---|---|
| DiOC2(3) (D14730) DiOC5(3) (D272) DiOC6(3) (D273) DiSC3(5) (D306) DiIC1(5) (H14700) | Carbocyanine (cationic) | SLOW; fluorescence response to depolarization depends on the staining concentration and detection method | |
| JC-1 (T3168) JC-9 (D22421) | Carbocyanine (cationic) | SLOW; fluorescence emission ratio 585/520 nm increases upon membrane hyperpolarization | |
| Tetramethylrhodamine methyl and ethyl esters (T668, T669) Rhodamine 123 (R302, R22420) | Rhodamine (cationic) | SLOW; used to obtain unbiased images of potential-dependent dye distribution | |
| Oxonol V (O266) Oxonol VI (O267) | Oxonol (anionic) | SLOW; fluorescence decreases upon membrane hyperpolarization | |
| DiBAC4(3) (B438, B24570) DiBAC4(5) (B436) DiSBAC2(3) (B413) | Oxonol (anionic) | SLOW; fluorescence decreases upon membrane hyperpolarization | |
| Merocyanine 540 (M24571) | Merocyanine | FAST/SLOW (biphasic response) |
For Research Use Only. Not for use in diagnostic procedures.