Search
Extinction Coefficient (EC) *
| ||||
|---|---|---|---|---|
| Product |
350 nm
|
405 nm
|
488 nm
|
532 nm
|
| Qdot 525 nanocrystals | 710,000 | 360,000 | 130,000 | NA |
| Qdot 565 nanocrystals | 1,900,000 | 1,100,000 | 290,000 | 139,000 |
| Qdot 585 nanocrystals | 3,500,000 | 2,200,000 | 530,000 | 305,000 |
| Qdot 605 nanocrystals | 4,400,000 | 2,800,000 | 1,100,000 | 580,000 |
| Qdot 625 nanocrystals | 14,700,000 | 9,900,000 | 2,700,000 | 870,000 |
| Qdot 655 nanocrystals | 9,100,000 | 5,700,000 | 2,900,000 | 2,400,000 |
| Qdot 705 nanocrystals | 12,900,000 | 8,300,000 | 3,000,000 | 2,100,000 |
| Qdot 800 nanocrystals | 15,800,000 | 10,600,000 | 4,100,000 | 2,100,000 |
| * Molar extinction coefficient in cm-1M-1 determined at the corresponding wavelength. NA = Not applicable. | ||||
For Research Use Only. Not for use in diagnostic procedures.