Search
| Cat. No. | Label | Ex/Em (nm) | Notes | Size |
|---|---|---|---|---|
| F1176 | Fluorescein | 494/518 | Original green-fluorescent conjugate | 500 µg |
| B13422 | Alexa Fluor 488 | 495/519 | Brightest and most photostable green-fluorescent conjugate | 500 µg |
| T1175 | Tetramethylrhodamine | 553/577 | An extensively used red-orange–fluorescent conjugate | 500 µg |
| B35451 | Alexa Fluor 555 | 555/565 | Bright and photostable red-orange–fluorescent conjugate | 500 µg |
| B13423 | Alexa Fluor 594 | 590/617 | Excellent dye to combine with green-fluorescent probes | 500 µg |
| B35450 | Alexa Fluor 647 | 650/668 | Excellent dye to combine with green- and orange-fluorescent probes | 500 µg |
| B35452 | Alexa Fluor 680 | 679/702 | Excellent dye to combine with green-, orange-, and red-fluorescent probes | 500 µg |
| B1196 | Biotin-XX | NA | Visualized with labeled avidins and streptavidins (Molecular Probes' selection of avidin, streptavidin, NeutrAvidin and CaptAvidin conjugates—Table 7.9) | 500 µg |
| B1601 | Unlabeled | NA | Useful as a control, as well as for radioiodination and for preparation of new conjugates | 1 mg |
| NA = Not applicable. | ||||
For Research Use Only. Not for use in diagnostic procedures.