Search
| Level of Labeling | Amount of DNA within Range of Assay (ng) * |
|---|---|
| 1 biotin: 10 bases | 13–264 |
| 1 biotin: 50 bases | 66–1300 |
| 1 biotin: 100 bases | 132–2640 |
| 1 biotin: 200 bases | 264–5280 |
| * To convert ng of DNA to nanomoles or picomoles, use 330 g/mole for the average molecular weight of a single base. | |
For Research Use Only. Not for use in diagnostic procedures.