Search
| Product | Glycoprotein Stain | Kit Type | Cat. No. |
|---|---|---|---|
| Pro-Q Emerald 300 Glycoprotein Stain Kits | Pro-Q Emerald 300 stain, Ex/Em = 280/530 nm | Gel Stain Kit (includes SYPRO Ruby protein gel stain *) | P21855 |
| Gel and Blot Stain Kit (does not include a total-protein stain) | P21857 | ||
| Pro-Q Emerald 488 Glycoprotein Stain Kit | Pro-Q Emerald 488 stain, Ex/Em = 510/520 nm | Gel and Blot Stain Kit (does not include a total-protein stain) | P21875 |
| * See Protein Detection on Gels, Blots and Arrays—Section 9.3 for a description of the SYPRO Ruby protein gel stain. | |||
For Research Use Only. Not for use in diagnostic procedures.