Search
| Cat. No. | Application | Bead Size | Concentration | Number of Colors |
|---|---|---|---|---|
| F8890 | Blood flow determination | 10 µm | 3.6 × 106 beads/mL | 7 colors, 10 mL each |
| F8891 | Blood flow determination | 15 µm | 1.0 × 106 beads/mL | 7 colors, 10 mL each |
| F8892 | Blood flow determination | 15 µm | 1.0 × 106 beads/mL | 5 colors, 10 mL each |
| F21015 | Blood flow determination | 15 µm | 1.0 × 106 beads/mL | 4 colors, 10 mL each |
| F10720 | Microinjection/tracing | 0.04 µm | 5% solids | 4 colors, 1 mL each |
For Research Use Only. Not for use in diagnostic procedures.