Search
| Cat. No. | Phycobiliprotein | Molecular Weight | Absorption Max (nm) | EC (cm-1M-1) | Emission Max (nm) | Fluorescence QY |
|---|---|---|---|---|---|---|
| P800 | B-phycoerythrin | 240,000 | 546, 565 | 2,410,000 | 575 | 0.98 |
| P801 | R-phycoerythrin | 240,000 | 496, 546, 565 | 1,960,000 | 578 | 0.82 |
| A803, A819 | Allophycocyanin | 104,000 | 650 | 700,000 | 660 | 0.68 |
| EC = extinction coefficient. QY = quantum yield. | ||||||
For Research Use Only. Not for use in diagnostic procedures.