Search
| Organism Type | Human | Human colonic carcinoma cell line (HCA-7) isolated from a well differentiated mucoid adenocarcinoma of the colon and maintained in vitro. It spontaneously synthesizes HLA-DR which is mainly intracytoplasmic. (J Pathol. 1985 Aug;146(4):337-44.) |
| Tissues | Colon | |
| Phenotype | Adherent | |
| Primary | yes | |
| Application | Protein Expression |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||