Search
| Microspheres (Ex/Em) * |
0.02 µm
|
0.04 µm
|
0.1 µm
|
0.2 µm
|
0.5 µm
|
1.0 µm
|
2.0 µm
|
4.0 µm
|
|---|---|---|---|---|---|---|---|---|
| Carboxylate-Modified Microspheres | ||||||||
| Europium luminescent (340–370/610) | F20880 2 mL | F20881 2 mL | ||||||
| Platinum luminescent (390/650) | F20886 2 mL | |||||||
| Blue (365/415) | F8781 10 mL | F8805 10 mL | F8814 10 mL | F8824 2 mL | ||||
| Blue (350/440) | F8797 10 mL | F8815 10 mL | ||||||
| Blue-green (430/465) | F13080 ‡ 5 mL | |||||||
| Yellow-green (505/515) | F8787 10 mL | F8795 † 1 mL | F8803 10 mL | F8811 10 mL | F8813 10 mL | F8823 10 mL F13081 ‡ | F8827 2 mL | |
| Nile red (535/575) | F8784 10 mL | F8819 10 mL | F8825 2 mL | |||||
| Orange (540/560) | F8792 † 1 mL | F8800 10 mL | F8809 10 mL | F8820 10 mL F13082 ‡ | ||||
| Red-orange (565/580) | F8794 † 1 mL | |||||||
| Red (580/605) | F8786 10 mL | F8793 † 1 mL | F8801 10 mL | F8810 10 mL | F8812 10 mL | F8821 10 mL F13083 ‡ | F8826 2 mL | |
| Crimson (625/645) | F8782 2 mL | F8806 2 mL | F8816 2 mL | |||||
| Dark red (660/680) | F8783 2 mL | F8789 † 1 mL | F8807 | |||||
| Infrared (715/755) | F8799 1 mL | |||||||
| Sulfate Microspheres | ||||||||
| Blue (365/415) | F8854 2 mL | |||||||
| Yellow-green (505/515) | F8845 10 mL | F8848 10 mL | F8852 10 mL | F8853 2 mL | F8859 2 mL | |||
| Red (580/605) | F8851 10 mL | F8858 2 mL | ||||||
| Aldehyde–Sulfate Microspheres | ||||||||
| Yellow-green (505/515) | F8760 10 mL | F8762 10 mL | ||||||
| Amine-Modified Microspheres | ||||||||
| Yellow-green (505/515) | F8764 5 mL | F8765 5 mL | ||||||
| Red (580/605) | F8763 5 mL | |||||||
| * Excitation (Ex) and emission (Em) maxima in nm. FluoSpheres beads are supplied as aqueous suspensions containing 2% solids and 2 mM sodium azide, except for the 0.04 µm microspheres (†), which are supplied as aqueous suspensions containing 5% solids without preservatives; the 1 µm microspheres for tracer studies (‡), which are supplied as aqueous suspensions containing 1010 microspheres per mL and 0.02% thimerosal; and the europium luminescent microspheres, which are supplied as aqueous suspensions containing 0.5% solids and 2 mM sodium azide. All sizes fall within a narrow range as discussed in Microspheres—Section 6.5. Sizes indicated in the above tables are nominal and may vary from batch to batch. Actual sizes, as determined by electron microscopy, are specified on the product labels. | ||||||||
For Research Use Only. Not for use in diagnostic procedures.