Molecular Probes Goat Anti-Mouse Isotype-Specific Antibodies—Table 7.2
Learn more in The Molecular Probes® Handbook
Table 7.2 Molecular Probes goat anti-mouse isotype-specific antibodies.
Fluorophore |
Abs/Em *
|
Goat Anti-Mouse Isotype-Specific Antibodies †
|
|||
---|---|---|---|---|---|
IgG1 (γ1) | IgG2a (γ2a) | IgG2b (γ2b) | IgG3 (γ3) | ||
Alexa Fluor 350 | 346/442 | A21120 | A21130 | A21140 | |
Fluorescein | 494/518 | A10530 | |||
Alexa Fluor 488 | 495/519 | A21121 | A21131 | A21141 | A21151 |
Alexa Fluor 546 | 556/573 | A21123 | A21133 | A21143 | |
Alexa Fluor 555 | 555/565 | A21127 | A21137 | A21147 | |
R-phycoerythrin | 496, 546, 565/578 | P21129 | P21139 | P21149 | |
Alexa Fluor 568 | 578/603 | A21124 | A21134 | A21144 | |
Alexa Fluor 594 | 590/617 | A21125 | A21135 | A21145 | A21155 |
Alexa Fluor 633 | 632/647 | A21126 | A21136 | A21146 | |
Allophycocyanin | 650/660 | A10541 | A10686 | ||
Alexa Fluor 647 | 650/668 | A21240 | A21241 | A21242 | |
Alexa Fluor 680 | 679/702 | A31562 | A31563 | A31564 | |
Horseradish peroxidase | NA | A10551 | A10685 | ||
Biotin | NA | A10519 | |||
Unlabeled | NA | A10538 | |||
* Approximate absorption (Abs) and fluorescence emission (Em) maxima, in nm. † The Alexa Fluor goat anti-mouse isotype-specific antibodies have been cross-adsorbed against mouse IgM, IgA, pooled sera, purified human paraproteins and the other isotypes to minimize crossreactivity. NA = Not applicable. |
For Research Use Only. Not for use in diagnostic procedures.