Search
| Color (Ex/Em *) |
Size
| ||
|---|---|---|---|
| 0.04 µm | 0.2 µm | 1.0 µm | |
| Blue-green (430/465) | F13080 † | ||
| Yellow-green (505/515) | F8795 ‡ | F13081 † | |
| Orange (540/560) | F8792 ‡ | F13082 † | |
| Red-orange (565/580) | F8794 ‡ | ||
| Red (580/605) | F8793 ‡ | F13083 † | |
| Dark red (660/680) | F8789 ‡ | ||
| Europium (365/610) | F20880 § | F20881 § | |
| Platinum (390/650) | F20886 § | ||
| * Fluorescence or luminescence excitation and emission maxima, in nm. † 1 × 1010 beads/mL, 5 mL unit size. ‡ 5% solids, 1.0 mL unit size. § 5% solids, 0.4 mL unit size. § 0.5% solids, 2.0 mL unit size. | |||