Search
| Cat. No. | Size of Coverslip (mm) | Number of Wells per Coverslip | Well Dimensions | Depth | Volume per Well | Number of Coverslips per Insert |
|---|---|---|---|---|---|---|
| CultureWell multiwell cell culture systems * † | ||||||
| C24762 | 24 × 50 | 2 | 15 mm diameter | 1 mm | 250–400 µL | 4 |
| C24763 | 24 × 50 | 2 | 15 mm diameter | 2 mm | 300–500 µL | 4 |
| C24764 | 24 × 50 | 3 | 9.5 mm square | 1 mm | 300–500 µL | 4 |
| C24765 | 24 × 50 | 4 | 9 mm diameter | 1 mm | 50–100 µL | 4 |
| C24766 C24767 | 24 × 50 | 8 | 6 mm diameter | 1 mm | 15–30 µL | 4 |
| C24768 | 24 × 50 | 50 | 3 mm diameter | 1 mm | 3–10 µL | 4 |
| CultureWell chambered coverslips ‡ | ||||||
| C24775 | 24 × 50 | 2 | 15 mm diameter | 1 mm | 250–400 µL | NA |
| C24776 | 24 × 50 | 2 | 15 mm diameter | 2 mm | 300–500 µL | NA |
| C24777 | 24 × 50 | 3 | 9.5 mm diameter | 1 mm | 300–500 µL | NA |
| C24778 | 24 × 50 | 4 | 9 mm diameter | 1 mm | 50–100 µL | NA |
| C24779 | 24 × 50 | 8 | 6 mm diameter | 1 mm | 15–30 µL | NA |
| C24780 | 24 × 50 | 50 | 3 mm diameter | 1 mm | 3–10 µL | NA |
| CultureWell multislip cell culture systems * | ||||||
| C24761 | 18 × 18 | NA | NA | NA | NA | 8 |
| C24760 | 12 × 12 | NA | NA | NA | NA | 15 |
| * Each system is supplied in packages of 10 inserts, each preassembled in an 86 mm × 128 mm plate. † A trial size is also available (C24767) that includes two inserts — four coverslips with eight 6 mm wells each — in two plates. ‡ Chambered coverslips are supplied in sets of coverslips, in five pouches of four coverslips each. Packs of 10 sterile plates are also available separately (C24769). NA = Not applicable. | ||||||
For Research Use Only. Not for use in diagnostic procedures.