Search
| Dye | Abs * | Extinction Coefficient † | Amine-Reactive Dyes ‡ | Notes |
|---|---|---|---|---|
| Dabcyl | 453 | 32,000 | D2245 (SE) |
|
| Malachite green | 628 | 76,000 | M689 (ITC) |
|
| QSY 7 | 560 | 90,000 | Q10193 (SE) |
|
| QSY 9 | 562 | 88,000 | Q20131 (SE) |
|
| QSY 21 | 661 | 90,000 | Q20132 (SE) |
|
| QSY 35 | 475 | 23,000 | Q20133 (SE) § |
|
| * Absorption (Abs) maxima, in nm. † Molar extinction coefficient in cm-1M-1 determined at the wavelength listed in the column headed Abs. These values may vary with the environment, particularly for the QSY 35 dye. ‡ (ITC) = Isothiocyanate. (SE) = Succinimidyl ester. | ||||
For Research Use Only. Not for use in diagnostic procedures.