Search
| Label(s) (Absorption/Emission Maxima) * | 3000 MW | 10,000 MW | 40,000 MW | 70,000 MW | 500,000 MW | 2,000,000 MW |
|---|---|---|---|---|---|---|
| Cascade Blue (400/420) | D7132 † | D1976 † | ||||
| Lucifer yellow (428/532) | D1825 † | |||||
| Alexa Fluor 488 (494/521) | D34682 | D22910 ‡ | ||||
| Oregon Green 488 (496/524) | D7170 D7171 † | D7172 D7173 † | ||||
| Fluorescein (494/518) | D3305 D3306 † | D1821 D1820 † | D1844 D1845 † | D1823 D1822 † | D7136 † | D7137 † |
| Fluorescein + biotin (494/518) | D7156 † | D7178 † | ||||
| Rhodamine Green (502/527) | D7163 | D7153 † | ||||
| BODIPY FL (505/513) | D7168 ‡ | |||||
| Oregon Green 514 (511/530) | D7176 | |||||
| Tetramethylrhodamine (555/580) | D3307 D3308 † | D1816 D1817 † D1868 ‡ | D1842 | D1819 D1818 † | D7139 † | |
| Tetramethylrhodamine + biotin (555/580) | D7162 † | D3312 † | ||||
| Alexa Fluor 555 (551/565) | D34679 ‡ | |||||
| Alexa Fluor 546 (556/573) | D22911 ‡ | |||||
| Rhodamine B (570/590) | D1824 | D1841 | ||||
| Alexa Fluor 568 (578/603) | D22912 ‡ | |||||
| Alexa Fluor 594 (590/617) | D22913 ‡ | |||||
| Texas Red (595/615) | D3329 D3328 † | D1828 D1863 † | D1829 | D1830 D1864 † | ||
| Alexa Fluor 647 (650/668) | D22914 ‡ | |||||
| Alexa Fluor 680 (681/704) | D34681 | D34680 ‡ | ||||
| Biotin (<300/none) | D7135 † | D1956 † | D1957 † | D7142 † | ||
| Amino (<300/none) | D3330 | D1860 | D1861 | D1862 | D7144 | |
| * Approximate absorption and emission maxima, in nm, for conjugates. † "Lysine-fixable" dextrans contain lysines and can therefore be fixed in place with formaldehyde or glutaraldehyde. ‡ "Fixable" dextrans contain free amines (but not lysines) and can be fixed in place with formaldehyde or glutaraldehyde. | ||||||
For Research Use Only. Not for use in diagnostic procedures.