Search
| Derivative | Abs * | Em * | Alkyl Halide or Haloacetamide | Maleimide | Other |
|---|---|---|---|---|---|
| Alexa Fluor 350 | 346 | 442 | A30505 | ||
| Alexa Fluor 405 | 402 | 421 | |||
| Anilinonaphthalene † | 326 | 462 | IAANS, I7 | MIANS, M8 | |
| Benzophenone | 282 | NA | B1508 | ||
| Bimane | 375 | 456 | B30500 | B30501 | |
| Dibromobimane | 394 | 490 | bBBr, D1379 ‡ | ||
| Diethylaminocoumarin | 384 | 470 | DCIA, D404 | CPM, D346 ‡ MDCC, D10253 | |
| Dimethylaminocoumarin | 376 | 465 | DACIA, D10252 | DACM, D10251 ‡ | |
| Dimethylaminonaphthalene † | 391 | 500 | badan, B6057 | acrylodan, A433 | |
| Monobromobimane | 394 | 490 | mBBr, M1378 ‡ mBBr, M20381 ‡ | ||
| Monochlorobimane | 394 | 490 | mBCl, M1381MP ‡ | ||
| Naphthalene † | 336 | 490 | IAEDANS, I14 | ||
| Pacific Blue | 410 | 455 | P30506 | ||
| Pacific Orange | 400 | 551 | P30507 | ||
| Phenanthroline | 270 | NA | P6879 | ||
| Pyrene † | 339 | 384 | P29 P2007MP | P28 | |
| Stilbene † | 329 | 408 | A484 | A485 | |
| * Approximate absorption (Abs) and emission (Em) maxima, in nm, for the reagent (if fluorescent) or the fluorescent thiol adduct. † Environment-sensitive fluorophore. ‡ Very weakly fluorescent until reacted with thiols. NA = Not applicable. | |||||
For Research Use Only. Not for use in diagnostic procedures.