Search
| Dyes (Cat #) | Structure (Charge) | Optical Response | Selected Applications |
|---|---|---|---|
| Di-4-ANEPPS (D1199) Di-8-ANEPPS (D3167) Di-2-ANEPEQ (D6923) Di-3-ANEPPDHQ (D36801) Di-4-ANEPPDHQ (D36802) | Styryl (cationic or zwitterionic) | FAST; fluorescence excitation ratio 440/505 nm decreases upon membrane hyperpolarization |
|
| RH 237 (S1109) RH 414 (T1111) RH 421 (S1108) RH 795 (R649) | Styryl (cationic or zwitterionic) | FAST; fluorescence decreases upon membrane depolarization |
For Research Use Only. Not for use in diagnostic procedures.