Search
| Donor |
Acceptor
| ||||
|---|---|---|---|---|---|
| QSY 35 | dabcyl | QSY 7 and QSY 9 | QSY 21 | ||
| Alexa Fluor 350 | 47 | 50 | |||
| Alexa Fluor 488 | 44 | 49 | 64 | ||
| Alexa Fluor 546 | 25 | 29 | 67 | ||
| Alexa Fluor 555 | 45 | ||||
| Alexa Fluor 568 | 56 | 75 | |||
| Alexa Fluor 594 | 77 | ||||
| Alexa Fluor 647 | 69 | ||||
| R0 values in angstroms (Å) represent the distance at which fluorescence resonance energy transfer from the donor dye to the acceptor dye is 50% efficient (Förster radius). Values were calculated from spectroscopic data as outlined (see Fluorescence Resonance Energy Transfer (FRET)—Note 1.2). | |||||