Search
| Target | Clone | Isotype | Qdot 565 | Qdot 605 | Qdot 655 | Qdot 705 | Qdot 800 |
|---|---|---|---|---|---|---|---|
| Human CD2 | S5.5 | Mouse IgG2a | Q10172 1 | Q221402 | Q221413 | Q22142 | |
| Human CD3 | UCHT1 | Mouse IgG2a | Q10054 1 | ||||
| Human CD3 | S4.1 | Mouse IgG2a | Q104842 Q100122 | Q22149 | |||
| Human CD4 | S3.5 | Mouse IgG2a | Q10480 1 Q10008 1 | Q104822 Q100072 | Q104853 Q100603 | Q22153 | |
| Human CD8 | 3B5 | Mouse IgG2a | Q10152 | Q10481 1 Q10009 1 | Q100552 | Q104833 Q100593 | Q22157 |
| Human CD10 | MEM-78 | Mouse IgG1 | Q10153 | Q22135 | Q22136 | Q10154 | |
| Human CD14 | TüK4 | Mouse IgG2a | Q10013 1 | Q100562 | Q221373 | Q10064 | |
| Human CD19 | SJ25-C1 | Mouse IgG1 | Q10306 | Q10179 | Q22138 | Q22139 | |
| Human CD20 | HI47 | Mouse IgG3 | Q10305 | ||||
| Human CD27 | CLB-27/1 | Mouse IgG2a | Q10065 1 | Q104862 Q100662 | |||
| Human CD38 | HIT2 | Mouse IgG1 | Q10053 | Q22150 Q10057 | Q22151 | Q22152 | |
| Human CD45 | HI30 | Mouse IgG1 | Q10051 | Q22154 | Q10062 | Q10156 | |
| Human CD45RA | MEM-56 | Mouse IgG2b | Q10047 | Q10069 | Q22155 | Q22156 | |
| Human CD56 | MEM-188 | Mouse IgG2b | Q10307 | ||||
| Human HLA-DR | Tü36 | Mouse IgG2b | Q10052 | Q22158 | Q22159 | Q10063 | |
| 1. Isotope control Q10014. 2. Isotype control Q10015. 3. Isotype control Q10076. All mouse anti–human CD antibody conjugates and isotype controls are supplied as 1 µM solutions in either 25 µL or 100 µL units. | |||||||
For Research Use Only. Not for use in diagnostic procedures.