Search
| Compound or Fluorophore | Azide or Alkyne | Cat. No. | Application |
|---|---|---|---|
| Azide- or Alkyne-Modified Biomolecules for Metabolic or Enzymatic Labeling | |||
| EdU (5-ethynyl-2'-deoxyuridine) | alkyne | A10044 E10187 E10415 | Detection of nascent DNA synthesis |
| EU (5-ethynyl uridine) | alkyne | E10345 | Detection of nascent RNA synthesis |
| Click-iT AHA (L-azidohomoalanine) | azide | C10102 | Detection of nascent protein synthesis or inhibition |
| Click-iT HPG (L-homopropargylglycine) | alkyne | C10186 | |
| Click-iT farnesyl alcohol, azide | azide | C10248 | Identification of isoprenylated proteins |
| Click-iT geranylgeranyl alcohol, azide | azide | C10249 | |
| Click-iT fucose alkyne (tetraacetylfucose alkyne) | alkyne | C10264 | Identification of fucosylated proteins |
| Click-iT LAA (linoleamide alkyne) | alkyne | C10447 | Identification of lipid peroxidation–derived protein modifications |
| Click-iT palmitic acid, azide | azide | C10265 | Identification of protein fatty acylation |
| Click-iT myristic acid, azide | azide | C10268 | |
| Click-iT GalNAz (tetraacetylated N-azidoacetylgalactosamine) | azide | C33365 | Identification of O-linked glycoproteins |
| Click-iT ManNAz (tetraacetylated N-azidoacetylmannosamine) | azide | C33366 | Identification of sialic acid–modified glycoproteins |
| Click-iT GlcNAz (tetraacetylated N-azidoacetylglucosamine) | azide | C33367 | Identification of O-linked glycoproteins |
| Azide- or Alkyne-Modified Fluorophores and Haptens for Detection (Ex/Em) * | |||
| Alexa Fluor 488 (495/519) | azide | A10266 | Fluorescent dye or hapten † |
| alkyne | A10267 | ||
| DIBO alkyne | C10405 | Fluorescent dye (or hapten †) for copper-free labeling and detection of azide-modified targets | |
| Alexa Fluor 555 (555/565) | azide | A20012 | Fluorescent dye |
| alkyne | A20013 | ||
| DIBO alkyne | C10406 | Fluorescent dye for copper-free labeling and detection of azide-modified targets | |
| Alexa Fluor 594 (590/617) | azide | A10270 | Fluorescent dye |
| alkyne | A10275 | ||
| DIBO alkyne | C10407 | Fluorescent dye for copper-free labeling and detection of azide-modified targets | |
| Alexa Fluor 647 (650/668) | azide | A10277 | Fluorescent dye |
| alkyne | A10278 | ||
| DIBO alkyne | C10408 | Fluorescent dye for copper-free labeling and detection of azide-modified targets | |
| Biotin (NA)‡ | azide | B10184 | Hapten |
| alkyne | B10185 | ||
| DIBO alkyne | C10412 | Hapten for copper-free labeling and detection of azide-modified targets | |
| Oregon Green 488 (496/524) | azide | O10180 | Fluorescent dye or hapten † |
| alkyne | O10181 | ||
| Tetramethylrhodamine (TAMRA) (555/580) | azide | T10182 | Fluorescent dye or hapten † |
| alkyne | T10183 | ||
| DIBO alkyne | C10410 | Fluorescent dye for copper-free labeling and detection of azide-modified targets | |
| * Absorption and fluorescence emission maxima, in nm. † As hapten, use with anti-dye antibody; see Anti-Dye and Anti-Hapten Antibodies—Section 7.4. ‡ Requires streptavidin for detection; see Avidin, Streptavidin, NeutrAvidin and CaptAvidin Biotin-Binding Proteins and Affinity Matrices—Section 7.6. | |||
For Research Use Only. Not for use in diagnostic procedures.