| Organism Type | Human | Species: human, Caucasian female 74 years old; Tissue: breast; Tumor: carcinoma
epithelial |
| Tissues | Breast/Mammary |
| Phenotype | Adherent |
| Primary | no |
| Application | Cell Culture / Growth Conditions,Transient Transfection |
Recommended Transfection Reagents
- Plasmid DNA transfection and co-transfection: Lipofectamine 3000 (>80% transfection efficiency and 3 fold protein expression improvement over Lipofectamine 2000)
- RNAi transfection (siRNA/miRNA) and gene silencing: Lipofectamine RNAiMAX
11875101,11965076,18324012,