Search
| Organism Type | Mouse | BALB/MK-2 cells are an epidermal growth factor (EGF)-dependent cell line derived from BALB/c mouse epidermis. Prototypical epithelial cell that can undergo differentiation to keratinocyte. |
| Tissues | Epithelial | |
| Phenotype | Adherent | |
| Primary | no | |
| Application | Transient Transfection |