Organism TypeHumanBronchial epithelial cells ATCC# CRL-9609 Isolated from normal human lung.
TissuesLung
PhenotypeAdherent
Primaryno
ApplicationCell Culture / Growth Conditions,Protein Expression,Transient Transfection,Virus Production