| Organism Type | Human | ATCC CCL-185; Properties: fatty acids synthesis; tumor markers; factor dependent; replication; enzymology; virology; lecithin synthesis; Susceptible to: adenovirus |
| Tissues | Lung | |
| Phenotype | Adherent | |
| Primary | no | |
| Application | Cell Culture / Growth Conditions,Nucleic Acid Purification,Protein Expression,Transfection: Gene Expression,Transient Transfection |
Recommended Transfection Reagents
- Plasmid DNA transfection and co-transfection: Lipofectamine® 3000 (51-79% transfection efficiency and 3 fold protein expression improvement over Lipofectamine® 2000)
- RNAi transfection (siRNA/miRNA) and gene silencing: Lipofectamine® RNAiMAX
Every step of the way, a wide range of cell health products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||