Search Thermo Fisher Scientific
Organism Type | Human | The Flp-In T-REx-293 cell line stably expresses the lacZ-Zeocin fusion gene and the Tet repressor, and is designed for use with the Flp-In T-REx System. The cell line expresses the Tet repressor from pcDNA6/TR and contains a single integrated Flp Recombination Target (FRT) site from pFRT/lacZeo as confirmed by Southern blot analysis. This cell line can be used as a host to generate a tetracycline-inducible Flp-In T-REx expression cell line by cotransfecting the pcDNA5/FRT/TO expression vector containing your gene of interest and the Flp recombinase expression plasmid, pOG44. |
Tissues | Kidney | |
Phenotype | Adherent | |
Primary | no | |
Application | Cell Culture / Growth Conditions,Protein Expression,Stable Cell Transfection,Transfection: Gene Expression |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||