Organism TypeHumanT-REx™ HeLa cells stably express the tetracycline repressor protein. All T-REx™ Cell Lines are functionally tested by transient transfection with the positive control vector pcDNA™4/TO/lacZ. T-REx™ Cell Lines exhibit extremely low basal expression levels.
TissuesCervix
PhenotypeAdherent
Primaryno
ApplicationCell Culture / Growth Conditions,Protein Expression,RNAi,Stable Cell Transfection,Transfection: Gene Silencing