Organism Type Primate - Non Human African Green Monkey
Tissues Kidney
Phenotype Adherent
Primary no
Application Cell Culture / Growth Conditions,Stable Cell Transfection,Transient Transfection,Virus Production
Recommended Transfection Reagents
  • Plasmid DNA transfection and co-transfection: Lipofectamine 3000 (30–50% transfection efficiency and 1 fold protein expression improvement over Lipofectamine 2000)
  • RNAi transfection (siRNA/miRNA) and gene silencing: Lipofectamine RNAiMAX
Every Step of the Way, a Wide Range of Cell Health Products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.