Search
| Organism Type | Human | ATCC CRL-1740; also called LNCaP.FGC. This cell was cloned from the left supraclavicular lymph node of a patient with metastatic prostate cancer. Grows as clusters, lightly adherent. |
| Tissues | Prostate | |
| Phenotype | Adherent | |
| Primary | no | |
| Application | Cell Culture / Growth Conditions,Stable Cell Transfection,Transfection: Gene Expression,Transient Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||