Search
| Organism Type | Rat | The MR1 cell line was derived from Fischer 344 rat embryo fibroblast cells that have been transfected with a gene causing immortality (c-myc). The MR1 line has been further transfected with a gene causing tumor formation (T24Ha-ras oncogene) |
| Tissues | Embryo | |
| Phenotype | Adherent | |
| Primary | no | |
| Application | Cell Culture / Growth Conditions |