Organism TypeHamsterATCC CCL-10; these cells are normally grown adherently, but some sublines have been adapted to suspension culture.
TissuesKidney
PhenotypeEither
Primaryno
ApplicationCell Culture / Growth Conditions,Protein Expression,Transient Transfection,Virus Production