Search
| Organism Type | Mouse | ATCC CRL-1934 and CRL-11632; pluripotent embryonic stem cells, full name is ES-D3 The cells spontaneously differentiate into embryonic structures in the absence of a feeder layer or conditioned medium. They can be maintained in the undifferentiated state by frequent subculture (every 2 to 3 days) on confluent feeder layers of irradiated (3000 rads) or mitomycin C treated (0.01 mg/ml for 90 minutes) primary mouse embryonic fibroblasts. Medium conditioned by Leukemia Inhibitory Factor (LIF) may also be used to maintain the undifferentiated state. LIF may also be added to ES-D3 cells cultured on feeder layers for additional support of the undifferentiated state. |
| Tissues | Embryo | |
| Phenotype | Adherent | |
| Primary | no | |
| Application | Cell Culture / Growth Conditions,Transient Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||