Search Thermo Fisher Scientific
Organism Type | Mouse | B82 L cells were devoid of endogenous EGFR. The B82L cells stably express wild-type human EGFR. |
Tissues | Fibroblast | |
Phenotype | Adherent | |
Primary | no | |
Application | Cell Culture / Growth Conditions,Stable Cell Transfection,Transfection: Gene Expression |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||