Search
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| 2 | 405 | 450/50 | 405 | 445 | (in buffer) 1 | flow cytometry |
The eFluor 450 dye is a violet-excitable, organic dye that emits at 450 nm and has spectral and performance properties similar to Pacific Blue dye. It exhibits equal or better fluorescence intensity when compared to Horizon V500 dye or Pacific Blue dye in multicolor applications. Stability data shows eFluor 450 exhibits minimal loss of fluorescence, when cells are exposed to a formaldehyde fixative treatment for up to 24 hours.
We offer eFluor 450 dye conjugated to primary antibodies for use in flow cytometry.
Search eFluor 450 primary antibodies Search eFluor 450 secondary antibodies
Comparison of fluorescence intensity of eFluor 450 dye with Pacific Blue. Human peripheral blood cells (PBMCs) were stained with CD45RA Monoclonal Antibody (HI100), eFluor 450 (purple line) or CD45RA Monoclonal Antibody (MEM-56), Pacific Blue (blue dotted line).
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
A handy reference poster featuring the broad range of our available dyes and labeling reagents.
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
A tool for selecting the optimal fluorescent dyes for your Invitrogen EVOS cell imaging systems.
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.