Search
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| 5 | 647 | Cy5 | 652 | 670 | 5 (in antifade) | Microscopy |
Invitrogen Aluora dyes, including Aluora 647 dye, are a specialized organic dye technology specifically optimized for spatial biology and multiplex immunohistochemistry (IHC) applications. Aluora 647 dye is excited by the 647 nm laser and emits at 670 nm. This dye has exceptional brightness and photostability. To learn more about spatial imaging, visit our Spatial Biology Resource Center.
We offer Aluora 647 spatial amplification reagents for use in immunohistochemistry.
Human FFPE small intestine was incubated with 1:25,000 of PCNA Monoclonal Antibody (PC10) (Cat. No. 13-3900) then labeled with the reagents in the Aluora 647 HRP-Mouse Kit (Cat. No. A40001334) and a DAPI nuclear stain. The slides were mounted in ProLong Glass mountant. Images were captured with an Invitrogen EVOS M7000 instrument (see information about EVOS imaging systems).
Invitrogen Aluora dyes are a specialized organic dye technology specifically optimized for spatial biology and multiplex immunohistochemistry applications. These bright, photostable dyes are excellent for detecting extremely low-abundance targets. Additionally, Aluora dyes exhibit a distinct emission spectra fingerprint that can be spectrally unmixed for high plex imaging experiments on spatial imaging platforms such as the EVOS S1000 Spatial Imaging System.
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
A handy reference poster featuring the broad range of our available dyes and labeling reagents.
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
A tool for selecting the optimal fluorescent dyes for your Invitrogen EVOS cell imaging systems.
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.