Search
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| 3 | 561 | 780/60 | 552 | 755 | (in buffer) 4 | flow cytometry |
NovaFluor Yellow 755 dye is excited by the yellow-green laser (561 nm) and is optimized for use in cell surface applications. It can be used for both spectral and conventional flow cytometry. With its distinct emission spectrum, NovaFluor Yellow 755 fits in a spectral space that is currently only accessible using cross-laser excited tandem dyes, such as PE-Cyanine7. Replacing highly cross-laser excited dyes may reduce spread, allowing for better resolution of low expressed markers. NovaFluor Yellow 755 dye should generally be paired with moderately expressed antigens to minimize spectral spillover in complex multicolor panels.
The macromolecule-based NovaFluor Yellow 755 dye produces stable fluorescence, and stained samples retain their fluorescence intensity and spectral signature when stored at 4°C. Use NovaFluor dyes with CellBlox Plus Blocking Buffer to reduce background and to block non-specific binding of NovaFluor labels, PE and APC tandems observed with macrophages and monocytes.
Thermo Fisher Scientific offers NovaFluor dyes conjugated to primary antibodies for use in flow cytometry, as well as NovaFluor Antibody Conjugation Kits, NovaFluor CD4 Label Characterization Kits, and custom conjugation services.
Spectral signature of NovaFluor Yellow 755 dye. Normal human peripheral blood cells stained with CD4 Monoclonal Antibody (SK3 (SK-3)), NovaFluor Yellow 755 were used for analysis. Data was acquired on a 5-laser Cytek Aurora system.
Staining of mouse splenocytes with NovaFluor Yellow 755 conjugated antibody. C57BL/6 mouse splenocytes were stained with CD3 Monoclonal Antibody (OKT3), Super Bright 436 only (left) or along with CD45 Monoclonal Antibody (HI30), NovaFluor Yellow 755 (right). Viable cells were used for analysis, as determined by LIVE/DEAD Fixable Blue Dead Cell Stain Kit, for UV excitation.
Staining of human PBMCs with NovaFluor Yellow 755 conjugated antibody. Normal human peripheral blood cells (PBMCs) were stained with CD19 Monoclonal Antibody (HIB19), eFluor 450 only (left) or with CD4 Monoclonal Antibody (SK3 (SK-3)), NovaFluor Yellow 755 (right). Cells in the lymphocyte gate were used for analysis.
NovaFluor dyes are built using Phiton technology and are compatible with both spectral flow cytometry and traditional flow cytometry. NovaFluor dyes exhibit narrow emission spectra and minimal cross-laser excitation, which helps reduce spectral spillover for better marker resolution. In addition, their unique spectral signatures can provide the opportunity to detect additional markers in flow cytometry panels by opening up previously unusable channels.
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
A handy reference poster featuring the broad range of our available dyes and labeling reagents.
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
A tool for selecting the optimal fluorescent dyes for your Invitrogen EVOS cell imaging systems.
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.