Search
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| 4 | 561 | 555/20 | 556 | 569 | (in buffer) 4 | flow cytometry |
Invitrogen Alexa Fluor 561 dye is a bright, yellow laser excitable dye that is optimized for flow cytometers with spectral capabilities. This fluorophore can be used for cell surface, intracellular, and intranuclear antigens. It should be used in complex multicolor panels to fit between fluorophores FITC and PE. In simpler panels, Alexa Fluor 561 can replace PE to reduce emission spreading into PE tandem dyes.
We offer Alexa Fluor 561 dye conjugated to primary antibodies for use in flow cytometry.
Search Alexa Fluor 561 primary antibodies Search Alexa Fluor 561 control antibodies
Flow cytometry analysis of C57BL/6 mouse splenocytes using CD4-Alexa Fluor 561. C57BL/6 mouse splenocytes were stained with CD45 Monoclonal Antibody, Alexa Fluor 532, CD3e Monoclonal Antibody, PE-Cyanine7, CD45R (B220) Monoclonal Antibody, PE, CD8a Monoclonal Antibody, PE-eFluor 610, CD25 Monoclonal Antibody, FITC and 0.06 µg of CD4 Monoclonal Antibody, Alexa Fluor 561. CD45 positive leukocytes were used for analysis and gated on CD3 positive/CD45R (B220) negative cells (left). CD3e positive cells were further plotted as CD4 versus CD8a positive cells (center) and CD4 positive cells were analyzed for expression of CD25 (right). This data was collected on a 5-laser Cytek Aurora full spectral cytometer. As shown here, Alexa Fluor 561 is compatible with PE and PE tandems when proper panel design principles are applied.
CD4-Alexa Fluor 561 non-specific staining in SJL mice. Balb/c mouse splenocytes (left) and SJL mouse splenocytes (right) were stained with Anti-Mouse CD19 Monoclonal Antibody, FITC and a non-cross reactive, human-specific CD4 Monoclonal Antibody (RPA-T4), Alexa Fluor 561. As shown here, Alexa Fluor 561-conjugated antibodies non-specifically stain B cells in SJL mice (right) and outbred, Swiss Webster mice (data not shown). Non-specific staining was not observed in BALB/c mice (left) and C57BL/6 mice (data not shown).
Protocols that fit your needs in flow cytometry ranging from sample preparation to numerous cell stimulation conditions, staining, immunophenotyping, and data analysis strategies.
A handy reference poster featuring the broad range of our available dyes and labeling reagents.
Protocols that fit your needs in imaging ranging from sample and assay preparation to staining, labeling, and data analysis strategies.
A tool for selecting the optimal fluorescent dyes for your Invitrogen EVOS cell imaging systems.
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.