Search Thermo Fisher Scientific
Organism Type | Rat | ATCC No.: CRL-1769 This line was originally isolated by D. Yaffe in 1969 from primary rat skeletal muscle cultures; no carcinogen was used to establish the L8 line; upon becoming confluent, L8 will fuse to form cross striated multinucleated muscle fibers |
Tissues | Myoblast | |
Phenotype | Adherent | |
Primary | yes | |
Application |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||