Search
Alexa Fluor and Alexa Fluor Plus Dyes |
Invitrogen Alexa Fluor and Alexa Fluor Plus dyes are widely used and trusted fluorescent dyes. These dyes are spectrally diverse, high-performing, small molecule fluorophores that are compatible with a variety of fluorescence-based applications. In addition to excellent performance, you'll benefit from an experienced, problem-solving technical support team, over 80,000 published references, and a wealth of application and experimental tips and protocols to aid in experimental planning.
With the wide range of Invitrogen Alexa Fluor and Alexa Fluor Plus dyes there is something for every filter and many options for multiplex detection. This series of fluorophores spans near-UV, visible and near-IR spectrum and are compatible with all common excitation sources and instruments.
| Color | Alexa Fluor and Alexa Fluor Plus dyes | Excitation/Emission | Dye equivalents |
|---|---|---|---|
| Blue | Alexa Fluor 350 | 343/441 | AMCA, coumarin |
| Alexa Fluor 405 | 401/422 | ||
| Alexa Fluor Plus 405 | 404/455 | ||
| Green | Alexa Fluor 430 | 431/540 | |
| Alexa Fluor 488 | 499/520 | Cy2, FITC (fluorescein) | |
| Alexa Fluor Plus 488 | 493/518 | ||
| Alexa Fluor 514 | 518/543 | ||
| Yellow | Alexa Fluor 532 | 534/553 | |
| Orange | Alexa Fluor 546 | 561/572 | |
| Alexa Fluor 555 | 553/568 | Cy3, TRITC (rhodamine) | |
| Alexa Fluor Plus 555 | 558/572 | ||
| Alexa Fluor 561 | 556/569 | ||
| Red | Alexa Fluor 568 | 579/603 | Rhodamine red |
| Alexa Fluor 594 | 590/618 | Texas Red | |
| Alexa Fluor Plus 594 | 590/618 | ||
| Far Red | Alexa Fluor 633 | 631/650 | |
| Alexa Fluor 635 | 633/648 | ||
| Alexa Fluor 647 | 650/671 | Cy5 | |
| Alexa Fluor Plus 647 | 658/675 | ||
| Near-IR | Alexa Fluor 660 | 663/691 | |
| Alexa Fluor 680 | 681/704 | Cy5.5, IR680 | |
| Alexa Fluor Plus 680 | 687/704 | ||
| Alexa Fluor 700 | 696/719 | ||
| Alexa Fluor 750 | 752/776 | Cy7 | |
| Alexa Fluor Plus 750 | 755/780 | ||
| Alexa Fluor 790 | 782/805 | ||
| Alexa Fluor Plus 800 | 789/794 |
Protein conjugates made with Alexa Fluor dyes produce fluorescence output that surpasses that of other spectrally similar fluorophore-labeled proteins. Alexa Fluor dyes are available in a wide selection of fluorescent colors and are more photostable than most other dyes.
Alexa Fluor dye molecules can be attached to proteins without significant self-quenching, leading to brighter conjugates available for imaging, flow cytometry, and other fluorescence-based applications.
Photostability is a key characteristic of Alexa Fluor dye conjugates, allowing more time for image capture. In addition to their exceptional brightness and the wide selection of dyes across the spectrum, you'll get photostability you can rely on with Alexa Fluor dyes and dye conjugates.
The photostability of Alexa Fluor dye molecules can be seen when compared to traditional fluorophores such as fluorescein.
Reduced photobleaching, sustained signal. Bovine pulmonary artery endothelial cells were labeled with fluorescein phalloidin (A) or Alexa Fluor 488 phalloidin (B) which label filamentous actin. The cells were placed under constant illumination on the microscope. Images were acquired at one-second intervals for 30 seconds. Under these illumination conditions, fluorescein images are photobleached to about 20% of its initial value, while fluorescence of Alexa Fluor 488 phalloidin stayed at the initial value.
While traditional Alexa Fluor dyes can be used in a variety of applications to detection proteins of varying expression levels, Alexa Fluor Plus dyes have been chemically optimized to provide higher signal to noise ratios and minimized cross-reactivity to help detect low-abundance targets in rare or precious samples.
Alexa Fluor Plus 405 conjugates allow for improved signal to noise when compared to traditional Alexa Fluor 405 conjugates. Traditional Alexa Fluor 405 secondary antibody (left) compared to Alexa Fluor Plus 405 secondary antibody (right, goat anti-rabbit IgG secondary antibody, Cat. No. A48254) in immunofluorescent analysis of β-III tubulin in fixed HeLa cells. Nuclei were stained with SYTOX Deep Red stain (Cat. No S11380). The images contain overlay of tubulin (cyan), and nuclei (magenta). Images were taken on an EVOS M7000 Imaging system (Cat. No. AMF7000) at 20x magnification.
For Research Use Only. Not for use in diagnostic procedures.