Helical scanning micro-computed X-ray tomography
Thermo Scientific HeliScan MicroCT brings a new era of micro computed tomography to materials science by leveraging advanced helical scanning and iterative reconstruction technology to produce excellent image fidelity. As part of a multi-scale imaging solution, HeliScan MicroCT enables you to gain valuable insight from internal structures to explore and validate a wide range of material properties.
HeliScan MicroCT features
Accurate segmentation
HeliScan MicroCT is the only microCT system specifically designed for quantification of high-quality, geometrically precise images.
Fast-throughput, high-quality analysis
Helical scanning and a large cone angle, combined with proprietary autofocus and drift correction, deliver high signal-to-noise ratio. Efficient scans of representative volumes provide ready-to-segment raw data that reduces the need for digital manipulation.
Better statistical relevance
Seamless representative volumes of high-aspect-ratio samples captured in one scan rather than inefficient, artifact-prone multi-scan with stitching.
Customizable, dynamic in situ experiments
Customizable feed-through ports, adaptable stages and a cabinet designed as a versatile, open platform easily support your experiments.
Ease of use and reliability
The system is optimized for multi-user environments, with advanced features available for expert users. Instrument components are chosen for dependability and serviceability.
Specifications
Style Sheet for Products Table Specifications
X-ray source
    20 - 160 kV, power = 8 W
X-ray detector
    3072 × 3072 pixels, 16 bit, flat pane
Spatial resolution
    800 nm
Sample diameter
    Up to 240 mm
Load capacity
    15 kg
Sample stage
  • Y-range: 
    • Source – Stage: 400 mm
    • Source – Detector: 750 mm
  • Z-range: 195 mm
  • R-range: 360° continuous
Style Sheet for Komodo Tabs
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards