Search
| Organism Type | Human | The MCF 10 cell line is a non-tumorigenic epithelial cell line. The line was produced by long term culture in serum free medium with low Ca++ concentration. MCF 10 was derived from adherent cells in the population. Cells derived from a floating population are available. The cells are positive for epithelial sialomucins, cytokeratins and milk fat globule antigen. They exhibit three dimensional growth in collagen, and form domes in confluent cultures. Thus far, the cells have shown no signs of terminal differentiation or senescence. The line is responsive to insulin, glucocorticoids, cholera endotoxin, and epidermal growth factor (EGF). By electron microscopy the cells display characteristics of luminal ductal cells but not of myoepithelial cells. They also express breast specific antigens as detected by positive reaction with MFA-Breast and MC-5 monoclonal antibodies. The calcium content of the medium exerts a strong effect on the morphology of the cells. |
| Tissues | Breast/Mammary | |
| Phenotype | Adherent | |
| Primary | no | |
| Application | Cell Culture / Growth Conditions |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||