Search
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. Thermo Fisher Scientific’s experienced team of GPCR biologists has generated a highly validated and novel family of GPCR cell line products to enable your discovery programs.
The portfolio contains two distinct assay platforms each utilizing FRET as the assay readout: Tango GPCR cell lines and GeneBLAzer GPCR cell lines. Each GPCR cell line platform is available in two cell states: Division Arrested non-dividing cells and normal dividing cells. Click through our cell line product table below and capability links to the right to see how we can help accelerate your drug discovery efforts.
| Assay Performance Data | Dividing Cells | Division Arrested Cells | Available in | ||||
|---|---|---|---|---|---|---|---|
| Family | Receptor | Geneblazer Technology | Tango Technology | Cat. No. | Cat. No. | SelectScreen Service | Confirmed Readout |
| Acetylcholine (Muscarinic) | M1 | K1710 | Ca²+(Fluo-4) | ||||
| Acetylcholine (Muscarinic) | M1 | K1735 | K1365 | ✓ | Ca²+(Fluo-4), Phospho-ERK2 | ||
| Acetylcholine (Muscarinic) | M1 | K1861 | Ca²+(Fluo-4), Phospho-ERK2 | ||||
| Acetylcholine (Muscarinic) | CHRM2 (M2) | K1564 | ✓ | Ca²+(Fluo-4) | |||
| Acetylcholine (Muscarinic) | M3 | K1716 | ✓ | Ca²+(Fluo-4) | |||
| Acetylcholine (Muscarinic) | M4 | K1741 | ✓ | Ca²+(Fluo-4) | |||
| Acetylcholine (Muscarinic) | M4 | K1818 | ✓ | Phospho-ERK2 | |||
| Acetylcholine (Muscarinic) | M5 | K1728 | ✓ | Ca²+(Fluo-4) | |||
| Adenosine | ADORA1 | K1445 | K1601 | ✓ | |||
| Adenosine | ADORA2A | K1530 | K1345 | cAMP | |||
| Adenosine | ADORA2B | K1780 | |||||
| Adenosine | ADORA3 | K1837 | |||||
| Adrenoreceptor | ADRA1A | K1470 | K1577 | ✓ | Ca²+(Fluo-4) | ||
| Adrenoreceptor | ADRA1B | K1471 | ✓ | Ca²+(Fluo-4) | |||
| Adrenoreceptor | ADRA2A | K1743 | ✓ | Ca²+(Fluo-4) | |||
| Adrenoreceptor | ADRA2A | K1604 | K1607 | ✓ | |||
| Adrenoreceptor | ADRB1 | K1553 | ✓ | cAMP | |||
| Adrenoreceptor | ADRB2 | K1472 | cAMP, ERK2 | ||||
| Adrenoreceptor | ADRB3 | K1745 | K1473 | ✓ | cAMP | ||
| Angiotensin | AGTR1 | K1443 | ✓ | Ca²+(Fluo-4), Phospho-ERK2 | |||
| Angiotensin-Like | AGTRL1 | K1754 | K1762 | ✓ | |||
| Bradykinin | BDKRB1 | K1734 | K1363 | ✓ | Ca²+(Fluo-4), Phospho-ERK2 | ||
| Bradykinin | BDKRB2 | K1790 | ✓ | ||||
| Calcitonin | CALCR | K1480 | ✓ | cAMP | |||
| Calcitonin | CALCRL: RAMP1 | K1437 | ✓ | cAMP | |||
| Calcitonin | CALCRL: RAMP2 | K1438 | cAMP | ||||
| Calcitonin | CALCRL: RAMP3 | K1439 | ✓ | cAMP | |||
| Calcium Sensing | CASR | K1833 | ✓ | Ca²+(Fluo-4) | |||
| Cannabinoid | CNR1 | K1513 | ✓ | ||||
| Cannabinoid | CNR2 | K1512 | K1567 | ✓ | |||
| Chemokine | CXCR1 | K1807 | ✓ | ||||
| Chemokine | CXCR2 | K1521 | ✓ | Ca²+(Fluo-4) | |||
| Chemokine | CXCR3 | K1829 | ✓ | Ca²+(Fluo-4) | |||
| Chemokine | CXCR4 | K1779 | ✓ | Ca²+(Fluo-4) | |||
| Chemokine | CXCR6 | K1563 | ✓ | ||||
| Chemokine | CXCR7 | K1832 | ✓ | ||||
| Chemokine | CCR1 | K1787 | K1793 | ✓ | Ca²+(Fluo-4) | ||
| Chemokine | CCR2 | K1809 | ✓ | ||||
| Chemokine | CCR3 | K1815 | ✓ | ||||
| Chemokine | CCR4 | K1799 | ✓ | Ca²+(Fluo-4) | |||
| Chemokine | CCR5 | K1788 | ✓ | Phospho-ERK2, Ca²+(Fluo-4) | |||
| Chemokine | CCR6 | K1761 | K1765 | ✓ | |||
| Chemokine | CCR7 | K1531 | ✓ | Phospho-ERK2, Ca²+(Fluo-4) | |||
| Chemokine | CMKLR1 | K1527 | ✓ | ||||
| Cholecystokinin | CCKAR | K1548 | K1623 | ✓ | Ca²+(Fluo-4), Phospho-ERK2 | ||
| Cholecystokinin | CCKBR | K1742 | K1379 | ✓ | Ca²+(Fluo-4) | ||
| Complement Component | C5AR1 | K1746 | K1554 | ✓ | Ca²+(Fluo-4) | ||
| Corticotropin Releasing Factor | CRHR1 | K1736 | K1367 | ✓ | cAMP | ||
| Corticotropin Releasing Factor | CRHR2 | K1733 | ✓ | cAMP | |||
| Dopamine | D1 | K1485 | cAMP | ||||
| Dopamine | D2 | K1708 | ✓ | Ca²+(Fluo-4), Phospho-ERK2 | |||
| Dopamine | D5 | K1784 | Ca²+(Fluo-4) | ||||
| Endothelial Differentiation Gene | EDG1 | K1520 | ✓ | ||||
| Endothelial Differentiation Gene | EDG2 | K1519 | ✓ | ||||
| Endothelial Differentiation Gene | EDG3 | K1713 | K1319 | ✓ | Ca²+(Fluo-4) | ||
| Endothelial Differentiation Gene | EDG3 | K1848 | Ca²+(Fluo-4) | ||||
| Endothelial Differentiation Gene | EDG4 | K1442 | ✓ | ||||
| Endothelial Differentiation Gene | EDG6 | K1622B | K1757 | ✓ | |||
| Endothelial Differentiation Gene | EDG7 | | K1511 | K1315 | ✓ | Ca²+(Fluo-4) | |
| Endothelial Differentiation Gene | EDG7 | | K1849 | ✓ | Ca²+(Fluo-4) | ||
| Endothelial Differentiation Gene | EDG8 | K1518 | ✓ | ||||
| Endothelin | EDNRA | K1726 | K1347 | ✓ | Ca²+(Fluo-4) | ||
| Endothelin | EDNRA | K1444 | ✓ | ||||
| Endothelin | EDNRB | K1727 | K1349 | ✓ | Ca²+(Fluo-4) | ||
| Formyl Peptide-Like | FPRL-1 | K1532 | K1357 | ✓ | Ca²+(Fluo-4) | ||
| Galanin | GALR1 | K1616 | ✓ | ||||
| Galanin | GALR2 | K1811 | ✓ | ||||
| Gastrin Releasing Peptide | GRPR | K1561 | K1637 | ✓ | |||
| Ghrelin | GHSR | K1819 | ✓ | Ca²+(Fluo-4) | |||
| Gastric Inhibitory Polypeptide | GIPR | K1864 | ✓ | ||||
| Glucagon | GCGR | K1855 | ✓ | cAMP | |||
| Glucagon | GLP1R | K1783 | ✓ | cAMP | |||
| Glucagon | GLP2R | K1543 | ✓ | cAMP | |||
| Gonadotropin Releasing Hormone | GnRHR | K1854 | ✓ | Ca²+(Fluo-4) | |||
| Histamine | H1 | K1703 | K1299 | ✓ | Ca²+(Fluo-4) | ||
| Histamine | H2 | K1707 | K1307 | ✓ | cAMP | ||
| Histamine | H3 | K1466 | K1509 | ✓ | |||
| Histamine | H4 | K1465 | ✓ | Ca²+(Fluo-4) | |||
| KiSS1 | GPR54 | K1720 | K1333 | ✓ | Ca²+(Fluo-4) | ||
| Leukotriene | LTB4R | K1748 | ✓ | Ca²+(Fluo-4) | |||
| Melanin-concentrating Hormone | MCHR1 (GPR24) | K1730 | ✓ | Ca²+(Fluo-4) | |||
| Melanin-concentrating Hormone | MCHR2 (GPR145) | K1852 | ✓ | Ca²+(Fluo-4) | |||
| Melanin-concentrating Hormone | MCHR2 (GPR145) | K1836 | ✓ | Ca²+(Fluo-4), Phospho-ERK2 | |||
| Melanocortin | MC1R | K1724 | ✓ | cAMP | |||
| Melanocortin | MC2R | K1483 | K1597 | ✓ | |||
| Melanocortin | MC3R | K1744 | ✓ | cAMP | |||
| Melanocortin | MC5R | K1740 | ✓ | cAMP | |||
| Melatonin | MTNR1A | K1669 | ✓ | ||||
| Melatonin | MTNR1B | K1753 | ✓ | ||||
| Motilin | MLNR | K1862 | |||||
| Neuropeptide B/W | GPR8 | K1478 | ✓ | ||||
| Neuropeptide S | NPSR1a | K1447 | K1493 | ✓ | Ca²+(Fluo-4) | ||
| Neuropeptide S | NPSR1b | K1481B | ✓ | Ca²+(Fluo-4) | |||
| Neuromedin U | NMUR1 | K1719 | ✓ | ||||
| Neurotensin | NTSR1 | K1781 | K1773 | ✓ | Ca²+(Fluo-4) | ||
| Neuropeptide Y | NPY1R | K1803 | ✓ | ||||
| Neuropeptide Y | NPY2R | K1479 | ✓ | ||||
| Neuropeptide Y | NPY5R | K1782 | |||||
| Nicotinic Acid | GPR109A | K1791 | ✓ | ||||
| Opioid | OPRD1 | | K1778 | ✓ | |||
| Opioid | OPRM1 | | K1523 | K1599 | ✓ | ||
| Opioid | OPRK1 | | K1533 | ✓ | Ca²+(Fluo-4). Phospho-ERK2 | ||
| Opioid | OPRK1 | | K1576 | ✓ | Phospho-ERK2 | ||
| Opioid | OPRL1 | | K1714 | ✓ | Phospho-ERK2 | ||
| Opioid | OPRL1 | | K1786 | ✓ | Ca²+(Fluo-4), Phospho-ERK2 | ||
| Orexin | HCRTR1 | | K1446 | ✓ | Ca²+(Fluo-4) | ||
| Orexin | HCRTR2 | | K1737 | K1369 | ✓ | Ca²+(Fluo-4) | |
| Platelet Activating Factor | PTAFR | | K1789 | ✓ | Ca²+(Fluo-4) | ||
| Prokineticin | PROKR2 | K1856 | |||||
| Prolactin Releasing Peptide | GPR10 | | K1732 | ✓ | Ca²+(Fluo-4) | ||
| Prostanoid | PTGDR | | K1739 | K1373 | ✓ | cAMP | |
| Prostanoid | CRTH2 (GPR44) | K1859 | ✓ | ||||
| Prostanoid | PTGER1 | K1851 | ✓ | ||||
| Prostanoid | PTGER2 | | K1723 | ✓ | cAMP | ||
| Prostanoid | PTGIR | K1850 | ✓ | ||||
| Prostanoid | TBXA2R | | K1549 | ✓ | Ca²+(Fluo-4), Phospho-ERK2 | ||
| Protease Activated | F2RL1 | | K1830 | ✓ | |||
| P2Y (Purinergic/ Pyrimidinergic) | P2RY2 | | K1722 | K1337 | ✓ | Ca²+(Fluo-4) | |
| P2Y (Purinergic) | P2RY4 | K1863 | Ca²+(Fluo-4) | ||||
| P2Y (Purinergic) | P2RY6 | | K1717 | K1327 | ✓ | ||
| P2Y (Purinergic) | P2RY11 | | K1729 | ✓ | Ca²+(Fluo-4) | ||
| P2Y (Purinergic) | P2RY14 | K1858 | |||||
| Relaxin | RLN3R1 | | K1569 | ✓ | |||
| Secretin | SCTR | | K1805 | ✓ | |||
| Serotonin | HTR1A (5HT1A) | K1857 | ✓ | Phospho-ERK2 | |||
| Serotonin | HTR2B | K1847 | ✓ | ||||
| Serotonin | HTR1D | | K1524 | ||||
| Serotonin | HTR2A | | K1562 | K1613 | ✓ | Ca²+(Fluo-4) | |
| Serotonin | HTR2C | K1877 | Ca²+(Fluo-4) | ||||
| Serotonin | HTR2C | Ca²+(Fluo-4) | |||||
| Serotonin | HTR7 | | K1704 | ✓ | |||
| Somatostatin | SSTR1 | | K1570 | ✓ | |||
| Somatostatin | SSTR2 | | K1582 | ✓ | |||
| Somatostatin | SSTR5 | | K1575 | K1573 | ✓ | ||
| Tachykinin | TACR1 | K1801 | ✓ | ||||
| Tachykinin | TACR2 | K1844 | ✓ | Ca²+(Fluo-4), Phospho-ERK2 | |||
| Tachykinin | TACR3 | | K1603 | ✓ | |||
| Vasopressin & Oxytocin | AVPR1a | | K1715 | K1323 | ✓ | Ca²+(Fluo-4), Phospho-ERK2 | |
| Vasopressin & Oxytocin | AVPR2 | | K1718 | K1329 | ✓ | cAMP | |
| Vasopressin & Oxytocin | OXTR | K1835 | ✓ | ||||
| VIP & PACAP | VPAC1 | | K1705 | ✓ | cAMP | ||
| VIP & PACAP | VPAC2 | | K1706 | ✓ | cAMP | ||
| VIP & PACAP | ADCYAP1R1 (PAC1) | | K1525 | K1619 | ✓ | cAMP | |
| Orphan | GPR1 | K1839 | cAMP | ||||
| Orphan | GPR21 | K1840 | |||||
| Orphan | GPR23 | K1841 | |||||
| Orphan | GPR35 | K1872 | ✓ | ||||
| Orphan | GPR39 | K1842 | |||||
| Orphan | GPR119 | | K1486 | K163B1 | ✓ | ||
| Orphan | GPR119 | | K1777 | K1770 | |||
| Orphan | GPR120 (short) | K1845 | |||||
| Orphan | GPR132 (G2A) | | K1749 | ||||
For Research Use Only. Not for use in diagnostic procedures.